Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
ACTTAGCCTAGCACCCAGCATATGGACAGTGCTAGGTTGTTGGGTTAAATCTCCCCTGATCCCACCCATCTTGTCCTTACTTCTGGCTGTATCTGCTTTGCCACAGATGGGAACACCCCATTCTTGTGTAGGAAGCAAATCTTTCAGCTTAGCAGGAAAGAAACATGTGATGTTTCTGCCACCCCTAGTTGAATGCTGTGCCCATCTCACAATAGGGCTTGATTTGGGTCATTCTATCTTCAACCACACACAATGGGCTCCTCTGAC
GenBank accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting mouse Ak036873 | description targeting human ZNF197-AS1 | description targeting human DIAPH3-AS2 | description targeting human AC018706 |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| esiRNA cDNA target sequence ACTTAGCCTAGCACCCAGCATATGGACAGTGCTAGGTTGTTGGGTTAAATCTCCCCTGATCCCACCCATCTTGTCCTTACTTCTGGCTGTATCTGCTTTGCCACAGATGGGAACACCCCATTCTTGTGTAGGAAGCAAATCTTTCAGCTTAGCAGGAAAGAAACATGTGATGTTTCTGCCACCCCTAGTTGAATGCTGTGCCCATCTCACAATAGGGCTTGATTTGGGTCATTCTATCTTCAACCACACACAATGGGCTCCTCTGAC | esiRNA cDNA target sequence CATGCCCCCAGCTAGTAAGAGGGTTCTATGTGTTGATTCACTGGGAAACCAGAGACTAGAGATACCTGTGAAGACACTTTCAAGAAGTTAGAAGAATTACCCTTGAATGAAGAAAGGAGCTGACTAGAAGTAGTTTATTGAATAACACACAAAGATTGTTTCAAACCTACAAAAGATAAATGTGAGGAAGGGATTTGGGGAATCCTCCTATGTCCTCCAGTCTGGC | esiRNA cDNA target sequence GAAGCCACACAATGCAGAGAATCAACAGTGGAGCTATGCCTAGTGAGAAGCAAAGAGACACTGGATGGCCTGAGAGGGAAAGAGACTGAGAAAAGCTGTCAGCTTTTAGTTCCTGGTTCCAGTCCTTTGTGACGGCCAGCTTGATTAGAGAGAAAAGGTCATTTGGAAAATATAGCCATTTTTGGAAGACTGAGCAGCCAAGGTTCTTAAGTATAGGAGATTGGGACAAAACAAAAAAAAGGTTTATAACAGTTCACTACTTCATCTACATCCTCAAGCCAACACCT | esiRNA cDNA target sequence TGGTTATGCAGCTTCCAGTGGACTCCATTGTTACCTGGTAATCATCAGTCAATTGCCATCACCCTTAACCTTAAAGGTGTCACTTTCGTACTTCAGGTATTCTTTCCAAAGTCCCTCTACCCCTTCTTCTCCCTGTCTTATCTGTCCCTGATCTCTGCCACCCTCAGTTGCTTACATTGGCCAACCAGCCTGAAATGGGAGAGGTTGAATCCTTCATGCTATGTGCATAGCCAACATTCCACAAGATTTATGTCACTAAAGCCTGTGAAAATGACAAATGGTGAGAAAGAGGGCCAAAAAGGGAAGCAAAAATAAATCCAACTACGATTTAACAAAGGGCGGCCCAGTATCTGTACCTAGTGGGTACTGTGATATCTTGAGGGACATGAATTTAACAAGAGAAATAAACTGCTGTATGAGGATCTTTAAAATGCTCTTGTTTTCCATTCAAATTCTCTTTAGTAAAGTTGCTTTTTGCCCTGAAGTCATAATCCAATTGTGTCAACGTGGTAATTAAACATCTGGATTGATTAGGTGGGGAA |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| NCBI accession no. | NCBI accession no. - | NCBI accession no. - | NCBI accession no. - |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| GenBank accession no. | GenBank accession no. | GenBank accession no. | GenBank accession no. |
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
