Skip to Content
Merck

Skip To

EMNC012211

MISSION® esiRNA

targeting mouse Ak051499

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGAAAGGTGGAACATGACATCCCAGCAGGAGAATTCTTCATGTTACCCTTCCCAGCCAATCCAATACCCAATTTTTCACCTACAGTCTGAAGAGTGGCAACAATGAATCTGCTCTCCATGAAAGGGCTGGAGAGATAGTTTAGCCTGTACAGTGCTTGCTACACAAGCTTGAGGACCTGAGTTCGGATCCCAGCAGTCATATCAAGAGCTGGCTACTTGTCACATGCCTATAGTTCCAGCTCCATGGGGGCATACACAGGGAGATCCCTGAAGCTCACTGACCAGTCACTCAAGCAGAACTGGTGAGCTCCAGGTTCAATGAGATAGCCTGACCCACCAA

GenBank accession no.

shipped in

ambient

storage temp.

−20°C

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EHNC011951EHNC010011EMNC008421
description

targeting mouse Ak051499

description

targeting human AL449106

description

targeting human AC016735.2

description

targeting mouse Ak053163

esiRNA cDNA target sequence

TGGAAAGGTGGAACATGACATCCCAGCAGGAGAATTCTTCATGTTACCCTTCCCAGCCAATCCAATACCCAATTTTTCACCTACAGTCTGAAGAGTGGCAACAATGAATCTGCTCTCCATGAAAGGGCTGGAGAGATAGTTTAGCCTGTACAGTGCTTGCTACACAAGCTTGAGGACCTGAGTTCGGATCCCAGCAGTCATATCAAGAGCTGGCTACTTGTCACATGCCTATAGTTCCAGCTCCATGGGGGCATACACAGGGAGATCCCTGAAGCTCACTGACCAGTCACTCAAGCAGAACTGGTGAGCTCCAGGTTCAATGAGATAGCCTGACCCACCAA

esiRNA cDNA target sequence

GGAATCTGTTGGTGGGAGAATTCCTGGATGTTTCCTACAAGCAGATGGCAATGTTCTCTTAGTCTCATCGGAACCAGACAGAGAAGGGGACGGTGGAAAAAGGACTGATAGAGACAAAGCACATCTAGCCAACCAGTCACCCTCTACTTCTAAAGACTTTTGAAAACAAGAGTATTGTCAAGAATTATTTAACATTATTATGATACTATCATTATAATCAAGGAGTGGGCTGTTGAGAGATTTCATCTTCATTCATTTTTTCAACAAATGTTTGGTCAGTGCCTAGGA

esiRNA cDNA target sequence

TTTGTGAGGATGGCTTTGTGTGGCTGCACATTGCCCTTGGCCAAGCCCCGCTGCCAGGAGAGGCAGCTCGGCTGCCAAACCTCCTGAACCCAAGTTACCTGAGCAAGCAGCCCTATCCCCTCGCCTGCTTCTCACTGCCTCGCCTGCCTTGGGCCTGCCCGACCCAGGGGGTCACATGCCTTGTTCCCCATCAGTGGAGACCAGGGACAAGAAGACAGTTCTGTGGGAAGAGCCAGGAGGACCATTTGGGAAAATACAGGTTCCAGGTGTGTCTCCAGACCTTGAGAGTTGGTGTCTCCTGGGAAGAGGCCCTGATGAGATCTGGGGAGATTCGGATCTGATGAGATTCAGGAAGCCCTGGCCTCTCCGTACTTGAGGAGACGGGACACACAGGACAAGCTGCAG

esiRNA cDNA target sequence

TCTGCTCGCAGTTGGTTATGTGGAGCCAGAAGGAAGTAGTGAACAGGCTCGTGCCTATAACCCCACCACTGGAGATGTGACAGCAGGGCAGTCACATCCAAGCCAACCTGTCTTGAGACCCTGTCTCAGAAATCTAAAAGCTAAAGGTGACATCTTAGAATGCCTCCCCCTGATGTGTTCCTTTGTAAATCCTGTGAAGATAGAAAACTCAAGACTCTCTCATTGCCTTGCTCAACAA

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

GenBank accession no.

AK051499

GenBank accession no.

AL449106

GenBank accession no.

AC016735

GenBank accession no.

AK053163

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters