Merck
  • Changes in volatile composition and sensory properties of Iranian ultrafiltered white cheese as affected by blends of Rhizomucor miehei protease or camel chymosin.
Changes in volatile composition and sensory properties of Iranian ultrafiltered white cheese as affected by blends of Rhizomucor miehei protease or camel chymosin.
Journal of dairy science (2016-07-18)
M Soltani, D Sahingil, Y Gokce, A A Hayaloglu
RESUMEN
The effect of using various combinations of Rhizomucor miehei protease and camel chymosin (100:0, 75:25, 50:50, 25:75, and 0:100, respectively) on volatile composition and sensory scores in Iranian ultrafiltered white cheese was studied during 90d of ripening. A solid-phase microextraction-gas chromatography-mass spectrometric method was used for determining the volatile compounds of the cheeses. Forty compounds including esters (12), acids (6), ketones (9), alcohols (3), and miscellaneous compounds (10) were identified. The main classes of volatile components in the cheeses are esters, miscellaneous compounds, and ketones. The type and concentration of the coagulants influenced both volatile composition and sensory scores of the cheeses. Principal component analysis separated the cheeses based on the use of 2 coagulants in various combinations and ripening time. The cheeses produced using higher concentrations of R. miehei were separately located on the plot compared with the cheeses produced using higher concentrations of camel chymosin. Sensory evaluation of the cheeses showed that, in general, the cheeses produced using higher concentrations of camel chymosin received higher body and texture and odor and flavor scores than the cheese produced using higher concentrations of R. miehei. In conclusion, 2 combinations of R. miehei and camel chymosin (75:25 and 25:75, respectively) can be successfully used for the production of Iranian ultrafiltered white cheese, considering the results of volatile composition and sensory analysis.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
Filtro SPME Polidimetilsiloxano (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
Filtro SPME Polidimetilsiloxano (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
Filtro SPME Polidimetilsiloxano (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME fiber assembly polyacrylate (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME fiber assembly polyacrylate (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
Kit de surtido de fibra SPME StableFlex, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
Filtro SPME Polidimetilsiloxano (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
Kit de surtido de fibra SPME StableFlex, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assortment kit 4, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, black hub