Merck
Inhibin Is a Novel Paracrine Factor for Tumor Angiogenesis and Metastasis.
Cancer research (2018-03-15)
Priyanka Singh, Laura M Jenkins, Ben Horst, Victoria Alers, Shrikant Pradhan, Prabhjot Kaur, Tapasya Srivastava, Nadine Hempel, Balázs Győrffy, Eugenia V Broude, Nam Y Lee, Karthikeyan Mythreye
RESUMEN
Inhibin is a heterodimeric TGFβ family ligand that is expressed in many cancers and is a selective biomarker for ovarian cancers; however, its tumor-specific functions remain unknown. Here, we demonstrate that the α subunit of inhibin (INHA), which is critical for the functionality of dimeric inhibin A/B, correlates with microvessel density in human ovarian tissues and is predictive of poor clinical outcomes in multiple cancers. We demonstrate that inhibin-regulated angiogenesis is necessary for metastasis. Although inhibin had no direct impact on tumor cell signaling, both tumor cell-derived and recombinant inhibin elicit a strong paracrine response from endothelial cells by triggering SMAD1/5 activation and angiogenesis in vitro and in vivo Inhibin-induced angiogenesis was abrogated via anti-inhibin α antibodies. The endothelial-specific TGFβ receptor complex comprising ALK1 and endoglin was a crucial mediator of inhibin signaling, offering a molecular mechanism for inhibin-mediated angiogenesis. These results are the first to define a role for inhibin in tumor metastasis and vascularization and offer an antibody-based approach for targeting inhibin therapeutically.Significance: Inhibin is a predictor of poor patient survival in multiple cancers and is a potential target for antiangiogenic therapies. Cancer Res; 78(11); 2978-89. ©2018 AACR.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-CD31 (PECAM-1)−FITC antibody produced in mouse, clone WM-59, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human ENG