Merck
DNA polymerase mu (Pol mu), homologous to TdT, could act as a DNA mutator in eukaryotic cells.
The EMBO journal (2000-04-04)
O Domínguez, J F Ruiz, T Laín de Lera, M García-Díaz, M A González, T Kirchhoff, C Martínez-A, A Bernad, L Blanco
RESUMEN
A novel DNA polymerase has been identified in human cells. Human DNA polymerase mu (Pol mu), consisting of 494 amino acids, has 41% identity to terminal deoxynucleotidyltransferase (TdT). Human Pol mu, overproduced in Escherichia coli in a soluble form and purified to homogeneity, displays intrinsic terminal deoxynucleotidyltransferase activity and a strong preference for activating Mn(2+) ions. Interestingly, unlike TdT, the catalytic efficiency of polymerization carried out by Pol mu was enhanced by the presence of a template strand. Using activating Mg(2+) ions, template-enhanced polymerization was also template-directed, leading to the preferred insertion of complementary nucleotides, although with low discrimination values. In the presence of Mn(2+) ions, template-enhanced polymerization produced a random insertion of nucleotides. Northern-blotting and in situ analysis showed a preferential expression of Pol mu mRNA in peripheral lymphoid tissues. Moreover, a large proportion of the human expressed sequence tags corresponding to Pol mu, present in the databases, derived from germinal center B cells. Therefore, Pol mu is a good candidate to be the mutator polymerase responsible for somatic hyper- mutation of immunoglobulin genes.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
ADN polimerasa Taq from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
ADN polimerasa Taq from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution