Merck
Estimating constants for metabolism of atrazine in freshly isolated rat hepatocytes by kinetic modeling.
Toxicology in vitro : an international journal published in association with BIBRA (2006-12-26)
Tami S McMullin, Melvin E Andersen, John D Tessari, Brian Cranmer, William H Hanneman
RESUMEN
This study estimated the kinetic constants for oxidative metabolism of atrazine (ATRA) and its chlorotriazine (Cl-TRI) metabolites, 2-chloro-4-ethylamino-6-amino-1,3,5-triazine (ETHYL), 2-chloro-4-amino-6-isopropylamino-1,3,5-triazine (ISO), and diaminochlorotriazine (DACT), using freshly isolated rat hepatocytes. Hepatocytes were incubated with 1.74, 44, 98, and 266 microM ATRA. Disappearance of ATRA and formation of the Cl-TRI metabolites were quantified over 90 min. At all incubation concentrations, ATRA was preferentially metabolized to ETHYL, producing ETHYL concentrations approximately 6 times higher than those of ISO. DACT concentrations peaked at 44 microM ATRA and decreased with increasing incubation concentrations, indicating non-linear metabolic behavior of ATRA with respect to DACT formation. A series of kinetic models were developed from these data to describe the dose and time-dependent oxidative metabolism of ATRA and the Cl-TRI metabolites. An integrated model for all the chloro-triazines included multi-substrate competitive inhibition of metabolism to describe the non-linear behavior of DACT production in relation to ATRA while simultaneously simulating the time-course behavior of the Cl-TRIs at all four ATRA concentrations. The maximal metabolic rate (V(max)) of ATRA metabolism and the Michaelis-Menten constant (K(M)) for the reaction were 1.6 microM/min and 30 microM, respectively. V(max) and K(M) values for ETHYL and ISO metabolism to DACT were also estimated using this modeling approach.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Trietazin, PESTANAL®, analytical standard