Merck
Lewis acid catalyzed propargylation of arenes with O-propargyl trichloroacetimidates: synthesis of 1,3-diarylpropynes.
The Journal of organic chemistry (2007-08-25)
Changkun Li, Jianbo Wang
RESUMEN
The BF3.OEt2-catalyzed Friedel-Crafts propargylation of aromatic compounds with O-propargyl trichloroacetimidates is highly efficient and affords 1,3-diarylpropyne derivatives in good yields.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Propyne, ≥97%
Sigma-Aldrich
Propyne, ≥99%