Merck
  • Effects of systemic administration of the essential oil of bergamot (BEO) on gross behaviour and EEG power spectra recorded from the rat hippocampus and cerebral cortex.
Effects of systemic administration of the essential oil of bergamot (BEO) on gross behaviour and EEG power spectra recorded from the rat hippocampus and cerebral cortex.
Functional neurology (2009-09-25)
Laura Rombolà, Maria Tiziana Corasaniti, Domenicantonio Rotiroti, Cristina Tassorelli, Shinobu Sakurada, G Bagetta, Luigi Antonio Morrone
RESUMEN
Bergamot (Citrus bergamia Risso et Poiteau) is a citrus fruit growing almost exclusively in the South of Italy. Its essential oil is obtained by cold pressing of the epicarp and, partly, of the mesocarp of the fresh fruit. Although this phytocomplex has been used for centuries, reputedly effectively, as a traditional medicine, there is very little verified scientific evidence to support this use. This paper reports original data on the systemic effects of the essential oil of bergamot (BEO) on gross behaviour and EEG activity recorded from the hippocampus and cerebral cortex of the rat. The Fast Fourier Transformation (FFT) was used to analyse and quantify the energy in single frequency bands of the EEG spectrum. The results obtained indicate that systemic administration of increasing volumes of BEO produces dose-dependent increases in locomotor and exploratory activity that correlate with a predominant increase in the energy in the faster frequency bands of the EEG spectrum. These data contribute to our understanding of the neurobiological profile of BEO.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bergamot oil