Merck
Enantioselective synthesis of planar chiral ferrocenes via palladium-catalyzed direct coupling with arylboronic acids.
Journal of the American Chemical Society (2012-12-21)
De-Wei Gao, Yan-Chao Shi, Qing Gu, Zheng-Le Zhao, Shu-Li You
RESUMEN
Enantioselective Pd(II)-catalyzed direct coupling of aminomethylferrocene derivatives with boronic acids was realized. With commercially available Boc-L-Val-OH as a ligand, planar-chiral ferrocenes could be synthesized in yields of 14-81% with up to 99% ee under mild conditions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ferrocene, 98%