Merck
  • A catalytic asymmetric ring-expansion reaction of isatins and α-alkyl-α-diazoesters: highly efficient synthesis of functionalized 2-quinolone derivatives.
A catalytic asymmetric ring-expansion reaction of isatins and α-alkyl-α-diazoesters: highly efficient synthesis of functionalized 2-quinolone derivatives.
Angewandte Chemie (International ed. in English) (2012-07-24)
Wei Li, Xiaohua Liu, Xiaoyu Hao, Yunfei Cai, Lili Lin, Xiaoming Feng
RESUMEN
Asymmetric expansion: A catalytic asymmetric ring-expansion reaction of the title compounds occurs in the presence of a Sc(OTf)(3) catalyst bearing an N,N'-dioxide-based ligand. Highly functionalized 2-quinolone derivatives containing a chiral C4-quaternary stereocenter were obtained in high yields and high levels of selectivity under mild reaction conditions (see scheme; Tf=trifluoromethanesulfonyl).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Scandium, powder, 99.9% trace rare earth metals basis