Merck
Triterpenic content and chemometric analysis of virgin olive oils from forty olive cultivars.
Journal of agricultural and food chemistry (2009-03-31)
Yosra Allouche, Antonio Jiménez, Marino Uceda, M Paz Aguilera, José Juan Gaforio, Gabriel Beltrán
RESUMEN
Forty olive cultivars (Olea europaea, L.) from the World Olive Germoplasm Bank Collection of Cordoba (Spain) were studied for their oil triterpenic dialcohol (uvaol and erythrodiol) and acid (oleanolic, ursolic, maslinic) composition. Dialcohol content ranged from 8.15 to 85.05 mg/kg, erythrodiol being the most predominant (from 5.89 to 73.78 mg/kg), whereas uvaol content was found at lower levels (from 1.50 to 19.35 mg/kg). Triterpenic acid concentration oscillated between 8.90 to 112.36 mg/kg. Among them, ursolic acid was found at trace levels, while the mean values of oleanolic and maslinic acids ranged from 3.39 to 78.83 mg/kg and 3.93 to 49.81 mg/kg, respectively. The variability observed for both triterpenic dialcohols and acid content was emphasized by principal component and cluster analyses. Both analyses were able to discriminate between oil samples, especially by erythrodiol, oleanolic acid, and maslinic acids. Regarding these results, we conclude that the virgin olive oil triterpenic fraction can be considered as a useful tool to characterize monovarietal virgin olive oil.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Erythrodiol, analytical standard