Merck
Pharmacological study on Alzheimer's drugs targeting calcium/calmodulin-dependent protein kinase II.
Journal of pharmacological sciences (2011-08-09)
Shigeki Moriguchi
RESUMEN
In the brain of Alzheimer's disease patients, down-regulation of both cholinergic and glutamatergic systems have been found and is thought to play an important role in impairment of cognition, learning, and memory. Nefiracetam is a pyrrolidine-related nootropic drug exhibiting various pharmacological actions such as a cognitive-enhancing effect. The present study was undertaken to elucidate mechanisms underlying the action of nefiracetam on glutamatergic receptors and intracellular protein kinases. N-Methyl-D-aspartate (NMDA)-evoked currents were recorded from rat cortical neurons in long-term cultured primary neurons using the whole-cell patch-clamp technique. NMDA-evoked currents were greatly and reversibly potentiated by bath application of nefiracetam, resulting in a bell-shaped dose-response curve. The maximum potentiation of 170% relative to the control was produced at 10 nM. Treatment with an inhibitor of the glycine binding site of the NMDA receptor, 7-chlorokynurenic acid, at 1 µM prevented augmentation of NMDA-evoked currents by nefiracetam. In rat hippocampal CA1 slices, field excitatory postsynaptic potentials were recorded by stimulation of Schaffer collateral/commissural pathways. Nefiracetam treatment significantly enhanced long-term potentiation (LTP) with the same bell-shaped dose-response curve. Furthermore, nefiracetam-induced LTP enhancement was closely associated with calcium/calmodulin-dependent protein kinase II (CaMKII) activation with concomitant increase in phosphorylation of AMPA-type glutamate receptor subunit 1 (GluA1) (Ser-831) as a postsynaptic CaMKII substrate. In conclusion, nefiracetam enhances NMDA-receptor function through stimulation of its glycine binding site and nefiracetam-induced CaMKII activation likely contributes to improvement of cognition, learning, and memory.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Nefiracetam, solid