Merck
  • Comparison of the toxicity of silver nanoparticles and silver ions on the growth of terrestrial plant model Arabidopsis thaliana.
Comparison of the toxicity of silver nanoparticles and silver ions on the growth of terrestrial plant model Arabidopsis thaliana.
Journal of environmental sciences (China) (2014-02-14)
Haifeng Qian, Xiaofeng Peng, Xiao Han, Jie Ren, Liwei Sun, Zhengwei Fu
RESUMEN
Silver nanoparticles (AgNPs) are one of the most widely used nanomaterials, but the mechanism of AgNP toxicity in terrestrial plants is still unclear. We compared the toxic effects of AgNPs and Ag+ on Arabidopsis thaliana at the physiological, ultrastructural and molecular levels. AgNPs did not affect seed germination; however, they showed stronger inhibitory effect on root elongation than Ag+. The results of transmission electron microscopy and metal content analysis showed that AgNPs could be accumulated in leaves. These absorbed AgNPs disrupted the thylakoid membrane structure and decreased chlorophyll content, which can inhibit plant growth. By comparison, a small amount of Ag+ was absorbed by seedlings, and it did not pronouncedly affect chloroplast structure and other metal ion absorption as AgNPs did. Compared with Ag+, AgNPs could alter the transcription of antioxidant and aquaporin genes, indicating that AgNPs changed the balance between the oxidant and antioxidant systems, and also affected the homeostasis of water and other small molecules within the plant body. All the data from physiological, ultrastructural and molecular levels suggest that AgNPs were more toxic than Ag+.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis