Merck
Determination of porphyrin carbon isotopic composition using gas chromatography-isotope ratio monitoring mass spectrometry.
Journal of chromatography. A (2001-01-12)
Z Yu, G Sheng, J Fu, P Peng
RESUMEN
Carbon isotopic compositions of aetio I occurring in the form of free-base, nickel, demetallation, dihydroxysilicon(IV) and bis(tert.-butyldimethylsiloxy)silicon(IV) [(tBDMSO)2Si(IV)] have shown that it has experienced no obvious isotope fractionation during the synthesis of [(tBDMSO)2Si(IV)] porphyrin from aetio I. Here, aetio I porphyrin species such as free-base, nickel, demetallated and dihydroxysilicon were analyzed by the conventional method, namely it is combusted in sealed system, and followed by isotope ratio monitoring mass spectrometric analysis. [(tBDMSO)2Si(IV)] aetio I was assayed by gas chromatography-isotope ratio monitoring mass spectrometry (GC-IRMS). A porphyrin mixture of [(tBDMSO)2Si(IV)] aetio I and octaethylporphyrin was also prepared. Their carbon isotopic compositions measured by GC-IRMS indicate that no isotope exchange took place between the porphyrins during the synthesis of [(tBDMSO)2Si(IV)] porphyrins. This method is employed for delta13C determination of geoporphyrins from the Maoming and Jianghan oil shales.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Hexachlorodisilane, 96%