Merck
Intra-arterial methylprednisolone for severe steroid refractory gastrointestinal graft-versus-host disease.
Pediatric blood & cancer (2014-07-24)
Kaljit S Bhuller, Manraj K S Heran, John K M Wu, S Rod Rassekh
RESUMEN
Acute graft versus host disease (GVHD) is a significant complication of bone marrow transplantation with approximately half of patients being refractory to steroids. There are numerous second-line systemic immunosuppressive treatments but the overall prognosis is poor and these therapies are associated with high mortality due to infection. An alternative approach to systemic treatment for GVHD is targeted delivery of immunosuppression. We present two pediatric cases with steroid-refractory gastrointestinal GVHD who clinically responded to intra-arterial steroid administration. We also review the literature regarding this treatment modality with a particular emphasis in children.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
USP
6α-Methylprednisolone, United States Pharmacopeia (USP) Reference Standard
6α-Methylprednisolone, European Pharmacopoeia (EP) Reference Standard