Merck
Correlation between portal vein anatomy and bile duct variation in 407 living liver donors.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2014-12-19)
K Takeishi, K Shirabe, Y Yoshida, Y Tsutsui, T Kurihara, K Kimura, S Itoh, N Harimoto, Y-I Yamashita, T Ikegami, T Yoshizumi, A Nishie, Y Maehara
RESUMEN
Our aim was to determine whether variant bile duct (BD) anatomy is associated with portal vein (PV) and/or hepatic artery (HA) anatomy. We examined the associations between BD anatomy and PV and/or HA anatomy in 407 living donor transplantation donors. We also examined whether the right posterior BD (RPBD) course was associated with the PV and/or HA anatomy. Variant PV, HA and BD anatomies were found in 11%, 25% and 25%, respectively, of 407 donors enrolled in this study. The presence of a variant BD was more frequently associated with a variant PV than with a normal PV (61% vs. 20%, p < 0.0001). By contrast, the presence of a variant HA was not associated with a variant BD. A supraportal RPBD was found in 357 donors (88%) and an infraportal RPBD was found in 50 donors (12%). An infraportal RPBD was significantly more common in donors with a variant PV than in donors with a normal PV (30% vs. 10%, p = 0.0004). Variant PV, but not variant HA, anatomies were frequently associated with variant BD anatomy. Additionally, an infraportal RPBD was more common in donors with a variant PV than in donors with a normal PV.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
N- Metil-D-glucamina, 99.0-100.5% (titration)
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
N- Metil-D-glucamina, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
N- Metil-D-glucamina, 99.0-100.5% dry basis, meets USP testing specifications
Supelco
N- Metil-D-glucamina, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
N- Metil-D-glucamina, European Pharmacopoeia (EP) Reference Standard