Merck
  • Human herpesvirus 6 reactivation before engraftment is strongly predictive of graft failure after double umbilical cord blood allogeneic stem cell transplantation in adults.
Human herpesvirus 6 reactivation before engraftment is strongly predictive of graft failure after double umbilical cord blood allogeneic stem cell transplantation in adults.
Experimental hematology (2014-07-30)
Amandine Le Bourgeois, Myriam Labopin, Thierry Guillaume, Jacques Delaunay, Yohann Foucher, Benoit Tessoulin, Florent Malard, Sameh Ayari, Pierre Peterlin, Sophie Derenne, Patricia Herry, Anne Cesbron, Katia Gagne, Laurence Lodé, Marina Illiaquer, Berthe-Marie Imbert-Marcille, Steven Le Gouill, Philippe Moreau, Mohamad Mohty, Patrice Chevallier
RESUMEN
Our main objective was to determine new factors associated with engraftment and single-unit predominance after double umbilical cord blood (UCB) allogeneic stem-cell transplantation. Engraftment occurred in 78% of cases in this retrospective study including 77 adult patients. Three-year overall survival, disease-free survival, relapse incidence, and nonrelapse mortality were 55 ± 6%, 44 ± 6%, 33 ± 5%, and 23 ± 4%, respectively. In multivariate analysis, Human herpesvirus 6 reactivation during aplasia (hazard ratio [HR] = 2.63; 95% confidence interval [CI]: 1.64-4.17; p < 0.001), younger recipient age (<53 years) (HR = 1.97; 95% CI: 1.16-3.35; p = 0.012), and lower human leukocyte antigen matching between the two units (3 of 6 or 4 of 6) (HR = 2.09; 95% confidence interval: 1.22-3.59; p = 0.013) were the three factors independently associated with graft failure. Also, factors independently predicting the losing UCB unit were younger age of the UCB unit (odds ratio [OR] = 1.01; 95% CI: 1-1.02; p = 0.035), lower CD34(+) cell dose contained in the UCB unit (≤ 0.8 × 10(5)/kg) (OR = 2.55; 95% CI: 1.05-6.16; p = 0.04), and presence of an ABO incompatibility between the UCB unit and the recipient (OR = 2.53; 95% CI: 1.15-5.53; p = 0.02). Thus, Human herpesvirus 6 reactivation during aplasia, lower unit-unit human leukocyte antigen matching, and younger UCB unit age, as new unfavorable predictive factors, may represent new parameters to take into account after double UCB allogeneic stem-cell transplantation in adults. These results need to be confirmed prospectively, as they may influence unit selections and patient outcomes.
MATERIALES
Número de producto
Marca
Descripción del producto
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
USP
Cytarabine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Clofarabine, ≥98% (HPLC)