Merck
Development of quantum dots-based biosensor towards on-farm detection of subclinical ketosis.
Biosensors & bioelectronics (2015-05-16)
Xuan Weng, Longyan Chen, Suresh Neethirajan, Todd Duffield
RESUMEN
Early detection of dairy animal health issues allows the producer or veterinarian to intervene before the animals' production levels, or even survival, is threatened. An increased concentration of β-hydroxybutyrate (βHBA) is a key biomarker for diagnosis of subclinical ketosis (SCK), and provides information on the health stress in cows well before any external symptoms are observable. In this study, quantum dots (QDs) modified with cofactor nicotinamide adenine dinucleotide (NAD(+)) were prepared for the sensing event, by which the βHBA concentration in the cow's blood and milk samples was determined via fluorescence analysis of the functionalized QDs. The detection was performed on a custom designed microfluidic platform combining with a low cost and miniaturized optical sensor. The sensing mechanism was first validated by a microplate reader method and then applied to the microfluidic platform. Standard βHBA solution, βHBA in blood and milk samples from cows were successfully measured by this novel technology with a detection limit at a level of 35 µM. Side by side comparison of the developed microfluidic biosensor with a commercial kit presented its good performance.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tetrahidrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahidrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
1,2-Dicloroetano, anhydrous, 99.8%
Sigma-Aldrich
Adenina, ≥99%
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Adenina, BioReagent, suitable for cell culture
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, ≥95% (HPLC)
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, ≥99%
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
Tetrahidrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
3-Aminophenylboronic acid monohydrate, 98%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
Adenina, BioReagent, suitable for plant cell culture, ≥99%