Product Name
Anti-Rabbit IgG - Tracy 690 from goat 1 mg/mL
Product Number
61908
Product Brand
sigma
CAS Number
Molecular Weight
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | Very Faint Blue to Very Dark Blue |
| APPEARANCE (FORM) | Liquid |
| EMISSION MAXIMUM | 715 ± 5 nm |
| REMARKS ON LUMINESCENCE | Ex. 695 nm in 0.1 M PBS pH 7.0 |
| PROTEIN CONTENT (280 NM) | 0.8 - 0.0 mg/ml |
| DYE-TO-PROTEIN RATIO (MOLAR) | 2.0 - 0.0 |
| SDS GEL ELECTROPHORESIS | CORRESPONDS TO REQUIREMENTS |
| REM. ON GELELECTROPHORESIS | UNCONJUGATED DYE ≤ 5 % OF TOTAL FLUORESCENCE |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |