Merck
  • Production of N-acetyl-D-neuraminic acid by use of an efficient spore surface display system.
Production of N-acetyl-D-neuraminic acid by use of an efficient spore surface display system.
Applied and environmental microbiology (2011-03-29)
Xiaoman Xu, Chao Gao, Xifeng Zhang, Bin Che, Cuiqing Ma, Jianhua Qiu, Fei Tao, Ping Xu
ABSTRACT
Production of N-acetyl-D-neuraminic acid (Neu5Ac) via biocatalysis is traditionally conducted using isolated enzymes or whole cells. The use of isolated enzymes is restricted by the time-consuming purification process, whereas the application of whole cells is limited by the permeability barrier presented by the microbial cell membrane. In this study, a novel type of biocatalyst, Neu5Ac aldolase presented on the surface of Bacillus subtilis spores, was used for the production of Neu5Ac. Under optimal conditions, Neu5Ac at a high concentration (54.7 g liter⁻¹) and a high yield (90.2%) was obtained under a 5-fold excess of pyruvate over N-acetyl-D-mannosamine. The novel biocatalyst system, which is able to express and immobilize the target enzyme simultaneously on the surface of B. subtilis spores, represents a suitable alternative for value-added chemical production.
Sigma-Aldrich
N-Acetylneuraminic Acid Aldolase from microorganisms, lyophilized powder, ≥20 units/mg protein (biuret)