Merck
  • Atom arrangement strategy for designing a turn-on 1H magnetic resonance probe: a dual activatable probe for multimodal detection of hypochlorite.
Atom arrangement strategy for designing a turn-on 1H magnetic resonance probe: a dual activatable probe for multimodal detection of hypochlorite.
Chemical communications (Cambridge, England) (2011-05-18)
Tomohiro Doura, Hiroshi Nonaka, Shinsuke Sando
ABSTRACT
The first dual activatable hypochlorite ((-)OCl)-sensing probe was developed, based on a new proof-of-concept design involving signal-activatable (1)H chemical probes using the triple-resonance NMR technique. The probe enabled fluorescence-(1)H MR dual turn-on detection of (-)OCl in solution and in crude tissue extracts.
Sigma-Aldrich
Dihydrocoumarin, 99%
Sigma-Aldrich
Dihydrocoumarin, ≥99%, FCC, FG