Skip to Content
Merck

JWPH3249C40F

J & W Pharmlab LLC

6-Formyl-indan-1-one 1,2-ethanediol ketal

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
204.23
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

2,3-dihydrospiro[indene-1,2'-[1,3]dioxolane]-6-carbaldehyde

Inchi Code

1S/C12H12O3/c13-8-9-1-2-10-3-4-12(11(10)7-9)14-5-6-15-12/h1-2,7-8H,3-6H2

InChI key

OFSSZQKGBNZZAD-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Brown Solid

Product Link

https://www.jwpharmlab.com/home/product/10R0179

MSDS

https://www.jwpharmlab.com/home/product/10R0179


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service