Merck
  • Staphylococcus aureus small colony variants impair host immunity by activating host cell glycolysis and inducing necroptosis.
Staphylococcus aureus small colony variants impair host immunity by activating host cell glycolysis and inducing necroptosis.
Nature microbiology (2019-11-07)
Tania Wong Fok Lung, Ian R Monk, Karen P Acker, Andre Mu, Nancy Wang, Sebastián A Riquelme, Silvia Pires, Loreani P Noguera, Felix Dach, Stanislaw J Gabryszewski, Benjamin P Howden, Alice Prince
ABSTRACT
Staphylococcus aureus small colony variants (SCVs) are frequently associated with chronic infection, yet they lack expression of many virulence determinants associated with the pathogenicity of wild-type strains. We found that both wild-type S. aureus and a ΔhemB SCV prototype potently activate glycolysis in host cells. Glycolysis and the generation of mitochondrial reactive oxygen species were sufficient to induce necroptosis, a caspase-independent mechanism of host cell death that failed to eradicate S. aureus and instead promoted ΔhemB SCV pathogenicity. To support ongoing glycolytic activity, the ΔhemB SCV induced over a 100-fold increase in the expression of fumC, which encodes an enzyme that catalyses the degradatin of fumarate, an inhibitor of glycolysis. Consistent with fumC-dependent depletion of local fumarate, the ΔhemB SCV failed to elicit trained immunity and protection from a secondary infectious challenge in the skin. The reliance of the S. aureus SCV population on glycolysis accounts for much of its role in the pathogenesis of S. aureus skin infection.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFRSF1A
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution