Merck
  • Immunogenicity of aggregates of recombinant human growth hormone in mouse models.
Immunogenicity of aggregates of recombinant human growth hormone in mouse models.
Journal of pharmaceutical sciences (2009-07-02)
Amber Haynes Fradkin, John F Carpenter, Theodore W Randolph
ABSTRACT
Aggregation of recombinant therapeutic protein products is a concern due to their potential to induce immune responses. We examined the immunogenicity of protein aggregates in commercial formulations of recombinant human growth hormone produced by freeze-thawing or agitation, two stresses commonly encountered during manufacturing, shipping and handling of therapeutic protein products. In addition, we subjected each preparation to high-pressure treatment to reduce the size and concentration of aggregates present in the samples. Aggregates existing in a commercial formulation, as well as aggregates induced by freeze-thawing and agitation stresses enhanced immunogenicity in one or more mouse models. The use of high-pressure treatment to reduce size and concentrations of aggregates within recombinant human growth hormone formulations reduced their overall immunogenicity in agreement with the "immunon" hypothesis.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) Alkaline Phosphatase conjugate, 1 mg/mL, Chemicon®
Pricing and availability is not currently available.