Merck
  • Induction of mammary tumors by expression of polyomavirus middle T oncogene: a transgenic mouse model for metastatic disease.
Induction of mammary tumors by expression of polyomavirus middle T oncogene: a transgenic mouse model for metastatic disease.
Molecular and cellular biology (1992-03-01)
C T Guy, R D Cardiff, W J Muller
ABSTRACT
The effect of mammary gland-specific expression of the polyomavirus middle T antigen was examined by establishing lines of transgenic mice that carry the middle T oncogene under the transcriptional control of the mouse mammary tumor virus promoter/enhancer. By contrast to most transgenic strains carrying activated oncogenes, expression of polyomavirus middle T antigen resulted in the widespread transformation of the mammary epithelium and the rapid production of multifocal mammary adenocarcinomas. Interestingly, the majority of the tumor-bearing transgenic mice developed secondary metastatic tumors in the lung. Taken together, these results suggest that middle T antigen acts as a potent oncogene in the mammary epithelium and that cells that express it possess an enhanced metastatic potential.
Sigma-Aldrich
AT-3 Mouse Mammary Carcinoma Cell Line, AT-3 mouse mammary tumor cell line may be used to develop mouse models for mammary cancer studies.
Pricing and availability is not currently available.