Merck
  • Alpha-actinin 4 potentiates myocyte enhancer factor-2 transcription activity by antagonizing histone deacetylase 7.
Alpha-actinin 4 potentiates myocyte enhancer factor-2 transcription activity by antagonizing histone deacetylase 7.
The Journal of biological chemistry (2006-09-19)
Sharmistha Chakraborty, Erin L Reineke, Minh Lam, Xiaofang Li, Yu Liu, Chengzhuo Gao, Simran Khurana, Hung-Ying Kao
ABSTRACT
Histone deacetylase 7 (HDAC7) is a member of class IIa HDACs that regulate myocyte enhancer factor-2 (MEF2)-mediated transcription and participate in multiple cellular processes such as T cell apoptosis. We have identified alpha-actinin 1 and 4 as class IIa HDAC-interacting proteins. The interaction domains are mapped to C terminus of alpha-actinin 4 and amino acids 72-172 of HDAC7. A point mutation in HDAC7 that disrupts its association with MEF2A also disrupts its association with alpha-actinin 4, indicating that MEF2A and alpha-actinin 4 binding sites largely overlap. We have also isolated a novel splice variant of alpha-actinin 4 that is predominantly localized in the nucleus, a pattern distinct from the full-length alpha-actinin 4, which is primarily distributed in the cytoplasm and plasma membrane. Using small interfering RNA, chromatin immunoprecipitation, and transient transfection assays, we show that alpha-actinin 4 potentiates expression of TAF55, a putative MEF2 target gene. Loss of MEF2A interaction correlates with loss of the ability of alpha-actinin 4 to potentiate TAF55 promoter activity. Ectopic expression of alpha-actinin 4, but not the mutant defective in MEF2A association, leads to disruption of HDAC7.MEF2A association and enhancement of MEF2-mediated transcription. Taken together, we have identified a novel mechanism by which HDAC7 activity is negatively regulated and uncovered a previously unknown function of alpha-actinin 4.
Sigma-Aldrich
HDAC10 FLAG tag active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥38% (SDS-PAGE)
Pricing and availability is not currently available.