Merck
  • Protein disulfide isomerase inhibition impairs Keap1/Nrf2 signaling and mitochondrial function and induces apoptosis in renal proximal tubular cells.
Protein disulfide isomerase inhibition impairs Keap1/Nrf2 signaling and mitochondrial function and induces apoptosis in renal proximal tubular cells.
American journal of physiology. Renal physiology (2020-08-25)
Indira D Pokkunuri, Mustafa F Lokhandwala, Anees Ahmad Banday
ABSTRACT
Renal proximal tubular apoptosis plays a critical role in kidney health and disease. However, cellular molecules that trigger renal apoptosis remain elusive. Here, we evaluated the effect of inhibiting protein disulfide isomerase (PDI), a critical thioredoxin chaperone protein, on apoptosis as well as the underlying mechanisms in human renal proximal tubular (HK2) cells. HK2 cells were transfected with PDI-specific siRNA in the absence and presence of an antioxidant, tempol. PDI siRNA transfection resulted in a decrease of ~70% in PDI protein expression and enzyme activity. PDI inhibition increased caspase-3 activity and induced profound cell apoptosis. Mitochondrial function, as assessed by mitochondrial cytochrome c levels, mitochondrial membrane potential, oxygen consumption, and ATP levels, was significantly reduced in PDI-inhibited cells. Also, PDI inhibition caused nuclear factor erythroid 2-related factor 2 (Nrf2; a redox-sensitive transcription factor) cytoplasmic sequestration, decreased superoxide dismutase and glutathione-S-transferase activities, and increased oxidative stress. In PDI-inhibited cells, tempol reduced apoptosis, caspase-3 activity, and oxidative stress and also restored Nrf2 nuclear translocation and mitochondrial function. Silencing Nrf2 in the cells abrogated the beneficial effect of tempol, whereas Kelch-like ECH-associated protein 1 (an Nrf2 regulatory protein) silencing protected cells from PDI inhibitory effects. Collectively, our data indicate that PDI inhibition diminishes Nrf2 nuclear translocation, causing oxidative stress that further triggers mitochondrial dysfunction and renal cell apoptosis. This study suggests an important role for PDI in renal cell apoptosis involving Nrf2 and mitochondrial dysfunction.
Sigma-Aldrich
MISSION® esiRNA, targeting human KEAP1
Pricing and availability is not currently available.