Merck
  • Lipase@zeolitic imidazolate framework ZIF-90: A highly stable and recyclable biocatalyst for the synthesis of fruity banana flavour.
Lipase@zeolitic imidazolate framework ZIF-90: A highly stable and recyclable biocatalyst for the synthesis of fruity banana flavour.
International journal of biological macromolecules (2020-11-09)
Tohid Taghizadeh, Atefeh Ameri, Amin Talebian-Kiakalaieh, Somayeh Mojtabavi, Alieh Ameri, Hamid Forootanfar, Sara Tarighi, Mohammad Ali Faramarzi
ABSTRACT
A zeolitic imidazolate framework (ZIF-90) has been synthesized through solvothermal method. The structure was characterized by means of FT-IR spectroscopy, X-ray diffraction, thermogravimetric analysis (TGA), and scanning electron microscopy (SEM)/energy dispersive X-ray spectroscopy (EDS). The synthesized ZIF-90 was applied as a support for immobilization of porcine pancreatic lipase (PPL). The immobilized enzyme (PPL@ZIF-90) exhibited immobilization yield and efficiency of 66 ± 1.8% and 89 ± 1.4%, respectively. The pH and thermal stability of PPL was improved after immobilization and the initial activity was retained at about 57% after 20 days of storage at 4 °C for PPL@ZIF-90. Moreover, about 57% of the original activity was remained following 10 cycles of application. In Michaelis-Menten kinetic studies, Km value for PPL@ZIF-90 was lower, while, the Vmax was higher than free PPL. Moreover, optimized conditions to produce fruity banana flavour upon esterification of butyric acid were investigated. The optimum esterification yield was 73.79 ± 1.31% in the presence of 245 mg PPL@ZIF-90, alcohol/acid ratio of 2.78 and 39 h reaction time. PPL@ZIF-90 showed 39% relative esterification yield after six cycles of reuse. The results suggested that PPL@ZIF-90 can be used as a potential effective biocatalyst for synthesis of isoamyl butyrate.
Sigma-Aldrich
2-Imidazolecarboxaldehyde, 97%
Pricing and availability is not currently available.