Merck
  • Spastin is a dual-function enzyme that severs microtubules and promotes their regrowth to increase the number and mass of microtubules.
Spastin is a dual-function enzyme that severs microtubules and promotes their regrowth to increase the number and mass of microtubules.
Proceedings of the National Academy of Sciences of the United States of America (2019-03-07)
Yin-Wei Kuo, Olivier Trottier, Mohammed Mahamdeh, Jonathon Howard
ABSTRACT
The remodeling of the microtubule cytoskeleton underlies dynamic cellular processes, such as mitosis, ciliogenesis, and neuronal morphogenesis. An important class of microtubule remodelers comprises the severases-spastin, katanin, and fidgetin-which cut microtubules into shorter fragments. While severing activity might be expected to break down the microtubule cytoskeleton, inhibiting these enzymes in vivo actually decreases, rather increases, the number of microtubules, suggesting that severases have a nucleation-like activity. To resolve this paradox, we reconstituted Drosophila spastin in a dynamic microtubule assay and discovered that it is a dual-function enzyme. In addition to its ATP-dependent severing activity, spastin is an ATP-independent regulator of microtubule dynamics that slows shrinkage and increases rescue. We observed that spastin accumulates at shrinking ends; this increase in spastin concentration may underlie the increase in rescue frequency and the slowdown in shortening. The changes in microtubule dynamics promote microtubule regrowth so that severed microtubule fragments grow, leading to an increase in the number and mass of microtubules. A mathematical model shows that spastin's effect on microtubule dynamics is essential for this nucleation-like activity: spastin switches microtubules into a state where the net flux of tubulin onto each polymer is positive, leading to the observed exponential increase in microtubule mass. This increase in the microtubule mass accounts for spastin's in vivo phenotypes.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin I antibody produced in mouse, clone SAP.4G5, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, Alexa Fluor 488 conjugate, clone DM1A, Upstate®, from mouse
Pricing and availability is not currently available.