Merck
  • Protein kinase C beta overexpression induces fibrotic effects in human proximal tubular epithelial cells.
Protein kinase C beta overexpression induces fibrotic effects in human proximal tubular epithelial cells.
The international journal of biochemistry & cell biology (2008-04-29)
Craig Slattery, Michael P Ryan, Tara McMorrow
ABSTRACT
Cyclosporine A (CsA) significantly improves the success of organ transplantation, however renal fibrosis, characterised by severe tubulointerstitial fibrosis is a complication of CsA therapy. Previously we have reported the involvement of PKC-beta isoforms in a model of CsA-induced tubulointerstitial fibrosis and we have now further elucidated this role. Treatment of human proximal tubular epithelial cells with CsA resulted in increased fibronectin production which coincided with increased PKC activity. To delineate the respective contributions of the two PKC-beta isoforms in fibrotic events, we overexpressed PKC-betaI, -betaII, or both in combination. Overexpression of the two PKC-beta isoforms induced morphological alterations, secretion of the profibrotic cytokine TGF-beta1, and fibronectin release from proximal tubular cells however PKC-betaII induced more significant effects in all parameters examined. Inhibition of PKC-beta completely abrogated the CsA-induced increase in fibronectin secretion demonstrating a direct antifibrotic effect of PKC-beta inhibition. Further studies also identified a role for the p44/42 mitogen activated kinase signalling pathway in CsA-induced fibrotic effects downstream of PKC-beta. Overall, these findings demonstrate a central role for PKC-beta, and particularly PKC-betaII in the development of tubulointerstitial fibrosis and suggest that PKC-beta may be a viable therapeutic target in CsA nephropathy.
Sigma-Aldrich
Hispidin, solid, ≥98% (HPLC)
Pricing and availability is not currently available.