Merck
  • Natural genetic variation in lycopene epsilon cyclase tapped for maize biofortification.
Natural genetic variation in lycopene epsilon cyclase tapped for maize biofortification.
Science (New York, N.Y.) (2008-01-19)
Carlos E Harjes, Torbert R Rocheford, Ling Bai, Thomas P Brutnell, Catherine Bermudez Kandianis, Stephen G Sowinski, Ann E Stapleton, Ratnakar Vallabhaneni, Mark Williams, Eleanore T Wurtzel, Jianbing Yan, Edward S Buckler
ABSTRACT
Dietary vitamin A deficiency causes eye disease in 40 million children each year and places 140 to 250 million at risk for health disorders. Many children in sub-Saharan Africa subsist on maize-based diets. Maize displays considerable natural variation for carotenoid composition, including vitamin A precursors alpha-carotene, beta-carotene, and beta-cryptoxanthin. Through association analysis, linkage mapping, expression analysis, and mutagenesis, we show that variation at the lycopene epsilon cyclase (lcyE) locus alters flux down alpha-carotene versus beta-carotene branches of the carotenoid pathway. Four natural lcyE polymorphisms explained 58% of the variation in these two branches and a threefold difference in provitamin A compounds. Selection of favorable lcyE alleles with inexpensive molecular markers will now enable developing-country breeders to more effectively produce maize grain with higher provitamin A levels.
Sigma-Aldrich
(±)-α-Carotene, ≥95.0% (HPLC)
Pricing and availability is not currently available.