Merck
  • Mitogen-activated protein kinase mediates mevalonate-stimulated human mesangial cell proliferation.
Mitogen-activated protein kinase mediates mevalonate-stimulated human mesangial cell proliferation.
Molecular medicine reports (2015-05-06)
Xiaoshuang Zhou, Chen Wang, Jihua Tian, Yanhong Wang, Yafeng Li, Zhaoyong Hu, Rongshan Li
ABSTRACT
The metabolic products of intracellular mevalonate (MVA) are important for the growth of eukaryotic cells. These products include cholesterol and several non-sterol isoprenoids. It has been reported that 3-hydroxy-3-methylglutaryl coenzyme A reductase inhibitors ameliorate glomerular injury in several experimental models of progressive glomerular disease by inhibiting the production of MVA and its metabolites. However, the mechanisms by which MVA stimulates the growth of human mesangial cells (HMCs) remain to be elucidated. To investigate the role of MVA in HMC proliferation, apoptosis, cell cycle and accumulation of extracellular matrix (ECM), the effects of MVA on HMCs at different durations and at various doses were evaluated. To examine the mechanisms of the effects of MVA on HMCs, the cells were treated with MVA, with or without PD98059, an extracellular signal-regulated kinase (ERK) inhibitor, SP600125, c-Jun NH2-teminal kinase (JNK) inhibitor, or SB203580, a P38 mitogen-activated protein kinase (MAPK) inhibitor. A 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide reduction assay was used to measure the proliferation of the HMCs, a flow cytometric assay was used to assess the proliferative index, and an ELISA was performed to determine the expression of transforming growth factor-β1 (TGF-β1), Type IV and Type I collagen (Col-IV and Col-I). The expression of B-cell lymphoma 2 (Bcl-2), Bcl-2-associated X protein (Bax), phosphorylated (p)-ERK1/2, p-JNK and p-p38 were also examined using western blot analysis. MVA significantly stimulated HMC proliferation and markedly increased the secretion of TGF-β1 and expression levels of Col-IV and Col-I. In addition, treatment with MVA significantly upregulated the expression of Bcl-2 and suppressed the expression of Bax in the HMCs. These responses were partially inhibited by the addition of inhibitors of ERK or JNK, however, they were not inhibited by the p38 MAPK inhibitor. These results demonstrated that MVA promoted HMC proliferation and ECM protein expression, which were associated with an increase in the expression of TGF-β1 and the inhibition of apoptosis. These effects were mediated, at least in part, by the JNK and ERK pathways.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free