- An improved procedure for the preparation of Ru(bpz)3(PF6)2 via a high-yielding synthesis of 2,2'-bipyrazine.
An improved procedure for the preparation of Ru(bpz)3(PF6)2 via a high-yielding synthesis of 2,2'-bipyrazine.
Beilstein journal of organic chemistry (2015-02-12)
Danielle M Schultz, James W Sawicki, Tehshik P Yoon
PMID25670993
ABSTRACT
There has been a recent surge of interest in the use of transition metal polypyridyl complexes as visible light-absorbing photocatalysts for synthetic applications. Among the most attractive features of this approach is the availability of many known complexes with well-characterized photophysical and electrochemical properties. In particular, Ru(bpz)3 (2+) is a powerful photooxidant that has proven to be uniquely suited for oxidatively induced photoredox transformations. We present here a straightforward and high-yielding route to Ru(bpz)3(PF6)2 that features an improved Pd-catalyzed synthesis of the 2,2'-bipyrazine ligand that is amenable to gram-scale preparations.