Merck
  • T-Cell Responses to Tyrosinase-Derived Self-Peptides in Patients with Leukoderma Induced by Rhododendrol: Implications for Immunotherapy Targeting Melanoma.
T-Cell Responses to Tyrosinase-Derived Self-Peptides in Patients with Leukoderma Induced by Rhododendrol: Implications for Immunotherapy Targeting Melanoma.
Dermatology (Basel, Switzerland) (2015-11-28)
Rie Takagi, Masaaki Kawano, Koichiro Nakamura, Tetsuya Tsuchida, Sho Matsushita
ABSTRACT
Rhododendrol, a phenolic compound contained in lightening/whitening cosmetics, can bind and inhibit tyrosinase and was reported to induce leukoderma in Japan. Only 2% of the cosmetics users are affected, and tacrolimus is effective in treatment of the condition. To test the hypothesis that the disease is an autoimmune disorder. Short-term T-cell lines were established using peripheral blood mononuclear cells from 8 patients with human melanoma-associated and tyrosinase-derived synthetic peptides. The effects of rhododendrol on melanoma immunization were also examined. Seven out of 8 patients were positive for HLA-DR4. Both class I- and class II-restricted and tyrosinase peptide-specific T-cell responses were observed. Immunization of mice with rhododendrol-treated and irradiated B16 melanoma cells successfully delayed the growth of melanoma cells in vivo. Rhododendrol-induced leukoderma is an autoimmune disorder, with rhododendrol as an environmental factor and HLA-DR4 as a genetic factor. Rhododendrol might be effective in treating melanomas.
Sigma-Aldrich
Rhododendrol, ≥95% (LC/MS-ELSD)
Pricing and availability is not currently available.