- Palladium-catalyzed selective 2,3-diarylation of alpha,alpha-disubstituted 3-thiophenemethanols via cleavage of C-H and C-C bonds.
Palladium-catalyzed selective 2,3-diarylation of alpha,alpha-disubstituted 3-thiophenemethanols via cleavage of C-H and C-C bonds.
The Journal of organic chemistry (2006-10-10)
Masaya Nakano, Tetsuya Satoh, Masahiro Miura
PMID17025334
ABSTRACT
Alpha,alpha-disubstituted 3-thiophenemethanols undergo selective diarylation accompanied by cleavage of the C-H and C-C bonds of the 2- and 3-positions, respectively, upon treatment with aryl bromides in the presence of a palladium catalyst to give the corresponding 2,3-diarylthiophenes in good yields.