Merck
  • Induction of high tolerance to artemisinin by sub-lethal administration: A new in vitro model of P. falciparum.
Induction of high tolerance to artemisinin by sub-lethal administration: A new in vitro model of P. falciparum.
PloS one (2018-01-18)
Serena De Lucia, Ioannis Tsamesidis, Maria Carmina Pau, Kristina R Kesely, Antonella Pantaleo, Francesco Turrini
ABSTRACT
Artemisinin resistance is a major threat to malaria control efforts. Resistance is characterized by an increase in the Plasmodium falciparum parasite clearance half-life following treatment with artemisinin-based combination therapies (ACTs) and an increase in the percentage of surviving parasites. The remarkably short blood half-life of artemisinin derivatives may contribute to drug-resistance, possibly through factors including sub-lethal plasma concentrations and inadequate exposure. Here we selected for a new strain of artemisinin resistant parasites, termed the artemisinin resistant strain 1 (ARS1), by treating P. falciparum Palo Alto (PA) cultures with sub-lethal concentrations of dihydroartemisinin (DHA). The resistance phenotype was maintained for over 1 year through monthly maintenance treatments with low doses of 2.5 nM DHA. There was a moderate increase in the DHA IC50 in ARS1 when compared with parental strain PA after 72 h of drug exposure (from 0.68 nM to 2 nM DHA). In addition, ARS1 survived treatment physiologically relevant DHA concentrations (700 nM) observed in patients. Furthermore, we confirmed a lack of cross-resistance against a panel of antimalarials commonly used as partner drugs in ACTs. Finally, ARS1 did not contain Pfk13 propeller domain mutations associated with ART resistance in the Greater Mekong Region. With a stable growth rate, ARS1 represents a valuable tool for the development of new antimalarial compounds and studies to further elucidate the mechanisms of ART resistance.
Sigma-Aldrich
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Pricing and availability is not currently available.