Skip to Content
Merck

692816

3-Iodo-2-methoxypyridine

97%

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C6H6INO
CAS Number:
Molecular Weight:
235.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

97%

form

liquid

refractive index

n20/D 1.618

density

1.831 g/mL at 25 °C

functional group

iodo

storage temp.

2-8°C

SMILES string

COc1ncccc1I

InChI

1S/C6H6INO/c1-9-6-5(7)3-2-4-8-6/h2-4H,1H3

InChI key

BXCHJERCAUZLOE-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service