Skip to Content
Merck

NIST3162A

Titanium (Ti) standard solution

NIST® SRM® 3162a

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

form

solution

packaging

pkg of 50 mL

application(s)

pharmaceutical (small molecule)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
NIST3155NIST3158NIST3167A
form

solution

form

-

form

-

form

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

-

packaging

pkg of 50 mL

packaging

pkg of 50 mL

packaging

pkg of 5 x 10 mL

packaging

pkg of 5 x 10 mL

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service