Skip to Content
Merck

Skip To

BPS60340

PDE4A1A Assay Kit

Synonym(s):

Assay Kits, PDE4A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41121811
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

technique(s)

activity assay: suitable (Fluorescence Polarization)

UniProt accession no.

shipped in

dry ice

storage temp.

-65 to -96°C

General description

The PDE4A Assay Kit is designed for identification of PDE4A inhibitors using fluorescence polarization. The assay is based on the binding of a fluorescent nucleotide monophosphate generated by PDE4A to the binding agent. The key to the PDE4A1A Assay Kit is the specific binding agent. Using this kit, only two simple steps on a microtiter plate are required for PDE4A1A reactions. First, the fluorescently labeled cAMP is incubated with a sample containing PDE4A1A for 1 hour. Second, a binding agent is added to the reaction mix to produce a change in fluorescent polarization that can then be measured using a fluorescence reader equipped for the measurement of fluorescence polarization.

Application

Study enzyme kinetics and screen small molecule inhibitors for drug discovery in high throughput screening (HTS) applications.
Phosphodiesterases (PDEs) play an important role in the dynamic regulation of the second messengers cAMP(cyclic adenosine monophosphate) and cGMP (cyclic guanosine monophosphate) signaling, by hydrolyzing them. The PDE superfamily is composed of 11 families, with PDE4, 7 and 8 being cAMP-specific hydrolases, and thus regulating positive and negative responses to it. PDE4 is the second most abundant PDE in cardiovascular tissues. PDE4, is a cAMP-specific PDE and the largest PDE subfamily with over 35 different isoforms and as such, the most widely characterized PDE isoenzyme. It is the predominant isoenzyme in the majority of inflammatory cells, and the airways smooth muscle and is implicated in inflammatory airways disease. PDE4 inhibitor roflumilast has been approved for the treatment of chronic obstructive lung disease.
Datasheet and Protocols may be found here.

Features and Benefits

The PDE4A1A inhibitor screening assay kit comes in a convenient 96-well format,including purified PDE4A1A enzyme, fluorescently labeled PDE4A substrate (cAMP),binding agent, and PDE assay buffer for 100 enzyme reactions.


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library