Skip to Content
Merck

213977

3,5-Diaminobenzyl alcohol dihydrochloride

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(H2N)2C6H3CH2OH·2HCl
CAS Number:
Molecular Weight:
211.09
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
248-868-1
MDL number:
Assay:
97%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

form

solid

mp

>300 °C (lit.)

functional group

hydroxyl

SMILES string

Cl.Cl.Nc1cc(N)cc(CO)c1

InChI

1S/C7H10N2O.2ClH/c8-6-1-5(4-10)2-7(9)3-6;;/h1-3,10H,4,8-9H2;2*1H

InChI key

JPEQTVYJBMDSFJ-UHFFFAOYSA-N

Application

3,5-Diaminobenzyl alcohol dihydrochloride has been used in the synthesis of :
  • 9-anilinothiazolo[5,4-b]quinoline derivatives, potential anti-tumor agents
  • {3-amino-5-[(7-chloro-4-quinolyl)amino]phenyl}methanol
  • luminescent lanthanide complexes consisting of a lanthanide-binding chelate and organic-based antenna molecule


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library