Skip to Content
Merck

Skip To

GF19655152

Copper - O.F.H.C.

foil, light tested, 50x50mm, thickness 0.075mm, half hard, 99.95+%

Synonym(s):

Copper, CV000540

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
1.010,00 PLN
5 ea
Please contact Customer Service for Availability
4.090,00 PLN
10 ea
Please contact Customer Service for Availability
7.470,00 PLN

About This Item

Linear Formula:
Cu
CAS Number:
Molecular Weight:
63.55
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141711
MDL number:
eCl@ss:
38150101

1.010,00 PLN


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 196-551-52

resistivity

1.673 μΩ-cm, 20°C

size × thickness

50 x 50 mm × 0.075 mm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF01396425GF61521320GF46192563
description

foil, light tested, 50x50mm, thickness 0.075mm, half hard, 99.95+%

description

foil, 10m coil, thickness 0.05mm, hard, 99.95+%

description

sheet, 50x50mm, thickness 6.35mm, 99.95+%

description

tube, 200mm, outside diameter 4.0mm, inside diameter 3mm, wall thickness 0.5mm, 99.95+%

form

foil

form

foil

form

wire

form

tubes

assay

99.95%

assay

99.95%

assay

99.95%

assay

99.95%

manufacturer/tradename

Goodfellow 196-551-52

manufacturer/tradename

Goodfellow 013-964-25

manufacturer/tradename

Goodfellow 615-213-20

manufacturer/tradename

Goodfellow 461-925-63

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters