Skip to Content
Merck

Skip To

GF29317439

Palladium

foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%

Synonym(s):

Palladium, PD000310

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 ea
Please contact Customer Service for Availability
17.140,00 PLN

About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141733
MDL number:
eCl@ss:
38191001

17.140,00 PLN


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 293-174-39

resistivity

9.96 μΩ-cm, 20°C

diam. × thickness

10 mm × 0.25 mm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

application(s)

catalysts

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF50988358GF97172556GF67339963
description

foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%

description

foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%

description

foil, 4mm disks, thickness 0.025mm, as rolled, 99.99+%

description

foil, 8mm disks, thickness 0.03mm, as rolled, 99.95%

form

foil

form

foil

form

foil

form

foil

assay

99.99%

assay

99.99%

assay

≥99.99%

assay

99.95%

manufacturer/tradename

Goodfellow 293-174-39

manufacturer/tradename

Goodfellow 509-883-58

manufacturer/tradename

Goodfellow 971-725-56

manufacturer/tradename

Goodfellow 673-399-63

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

mp

1554 °C (lit.)

mp

1554 °C (lit.)

mp

1554 °C (lit.)

mp

1554 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)


Still not finding the right product?

Explore all of our products under Palladium


Storage Class

13 - Non Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters