Skip to Content

BDPH9BCC1792

Bide Pharmatech

Ethyl 2-oxo-2-(phenylamino)acetate

Synonym(s):

ethyl anilino(oxo)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11O3N1
CAS Number:
Molecular Weight:
193.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H11NO3/c1-2-14-10(13)9(12)11-8-6-4-3-5-7-8/h3-7H,2H2,1H3,(H,11,12)

Inchi Key

YDGAUBHNAKCSKF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under Ethyl 2-oxo-2-(phenylamino)acetate


Signal Word

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library