Skip to Content
Merck

BDPH9BCC32FD

Bide Pharmatech

2-(Hydroxymethyl)benzonitrile

Synonym(s):

2-(hydroxymethyl)benzonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7O1N1
Molecular Weight:
133.15
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7NO/c9-5-7-3-1-2-4-8(7)6-10/h1-4,10H,6H2

Inchi Key

NWRXEGKWQXAEHC-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library