Skip to Content

BDPH9BCC3E13

Bide Pharmatech

3-Chloro-4-(pyridin-2-ylmethoxy)aniline

Synonym(s):

3-chloro-4-(2-pyridinylmethoxy)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H11O1N2Cl1
CAS Number:
Molecular Weight:
234.68
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H11ClN2O/c13-11-7-9(14)4-5-12(11)16-8-10-3-1-2-6-15-10/h1-7H,8,14H2

Inchi Key

XCAPJQSICQSUJP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Slightly pale reddish yellow - Yellow



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library