Skip to Content
Merck

BDPH9BCC4FAC

Bide Pharmatech

(3,5-Bis(trifluoromethyl)phenyl)(phenyl)methanone

Synonym(s):

[3,5-bis(trifluoromethyl)phenyl](phenyl)methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H8O1F6
CAS Number:
Molecular Weight:
318.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H8F6O/c16-14(17,18)11-6-10(7-12(8-11)15(19,20)21)13(22)9-4-2-1-3-5-9/h1-8H

Inchi Key

QETSMUIBVUASOJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid or lump



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library