Skip to Content
Merck

BDPH9BCC540D

Bide Pharmatech

5-Bromo-2-phenyl-1H-indole

Synonym(s):

5-bromo-2-phenyl-1H-indole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H10N1Br1
CAS Number:
Molecular Weight:
272.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H10BrN/c15-12-6-7-13-11(8-12)9-14(16-13)10-4-2-1-3-5-10/h1-9,16H

Inchi Key

XGOWHPSXPPZWAH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 5-Bromo-2-phenyl-1H-indole


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library