Skip to Content

BDPH9BCC613B

Bide Pharmatech

3-Bromo-6-(trifluoromethyl)pyridazine

Synonym(s):

3-bromo-6-(trifluoromethyl)pyridazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H2N2Br1F3
CAS Number:
Molecular Weight:
226.98
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H2BrF3N2/c6-4-2-1-3(10-11-4)5(7,8)9/h1-2H

Inchi Key

WCBUSZLXHSFLHK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Light yellow to Yellow Solid or Liquid



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library