Skip to Content
Merck

BDPH9BCC9042

Bide Pharmatech

1-(3,4-Dichlorophenyl)propan-1-amine

Synonym(s):

1-(3,4-dichlorophenyl)-1-propanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11N1Cl2
CAS Number:
Molecular Weight:
204.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11Cl2N/c1-2-9(12)6-3-4-7(10)8(11)5-6/h3-5,9H,2,12H2,1H3

Inchi Key

VBWRNUHVGFPJRG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library