Skip to Content
Merck

BDPH9BCC95B9

Bide Pharmatech

3,6-Dichlorobenzene-1,2-diamine

Synonym(s):

3,6-dichloro-1,2-benzenediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6N2Cl2
CAS Number:
Molecular Weight:
177.03
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6Cl2N2/c7-3-1-2-4(8)6(10)5(3)9/h1-2H,9-10H2

Inchi Key

GCGFESBSSZSYHD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 3,6-Dichlorobenzene-1,2-diamine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library